Product Details

SNP ID
rs17651424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112152903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATATATCAATTGTATGAATATGGA[C/T]GCAATATAATAACATAATTTCAAAT
Phenotype
MIM: 612141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPB41L4A PubMed Links

Gene Details

Gene
EPB41L4A
Gene Name
erythrocyte membrane protein band 4.1 like 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022140.3 Intron NP_071423.3
XM_011543530.1 Intron XP_011541832.1
XM_011543531.2 Intron XP_011541833.1
XM_011543532.1 Intron XP_011541834.1
XM_011543533.1 Intron XP_011541835.1
XM_017009689.1 Intron XP_016865178.1
Gene
EPB41L4A-AS1
Gene Name
EPB41L4A antisense RNA 1
There are no transcripts associated with this gene.

Gene
SNORA13
Gene Name
small nucleolar RNA, H/ACA box 13
There are no transcripts associated with this gene.

View Full Product Details