Product Details

SNP ID
rs17243975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161505388 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGCTGCTTCAGTAACGCCATCT[A/G]ATAGGTACATATCAAGTACTGCATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHCTF1P1 PubMed Links

Gene Details

Gene
AHCTF1P1
Gene Name
AT-hook containing transcription factor 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details