Product Details
- SNP ID
-
hCV43066661
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:43766430 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTTGTGTTTGACCATTTAAATA[C/G]TACACCTTTGAAATTTAAATAAAAA
- Phenotype
-
MIM: 109750
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BLVRA
PubMed Links
Gene Details
- Gene
- BLVRA
- Gene Name
- biliverdin reductase A
There are no transcripts associated with this gene.
View Full Product Details