Product Details

SNP ID
rs11539969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44796735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTACTATTAGCCATGGTCAACC[A/C]CACCGTGTTCTTCGACATTGCCGTC
Phenotype
MIM: 123840
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPIA PubMed Links

Gene Details

Gene
PPIA
Gene Name
peptidylprolyl isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300981.1 100 Intron NP_001287910.1
NM_021130.4 100 Missense Mutation CAC,CCC H4P NP_066953.1

View Full Product Details