Product Details

SNP ID
rs11558865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96231837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCTTCAAGTTCAGGTCCAGTGC[C/T]CTCTTAATGCGAAACATCCTGTCAT
Phenotype
MIM: 616930 MIM: 191330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTERF3 PubMed Links

Gene Details

Gene
MTERF3
Gene Name
mitochondrial transcription termination factor 3
There are no transcripts associated with this gene.

Gene
UQCRB
Gene Name
ubiquinol-cytochrome c reductase binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199975.2 413 Silent Mutation AGA,AGG R33R NP_001186904.1
NM_001254752.1 413 Silent Mutation AGA,AGG R65R NP_001241681.1
NM_006294.4 413 Silent Mutation AGA,AGG R65R NP_006285.1

View Full Product Details