Product Details

SNP ID
hCV43621381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58470965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATATTCAAAAACAGAAGAACAGG[G/T]GATATGATTGCTTCAGCAAGAAGCT
Phenotype
MIM: 614463
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NDRG4 PubMed Links

Gene Details

Gene
NDRG4
Gene Name
NDRG family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130487.1 Intron NP_001123959.1
NM_001242833.1 Intron NP_001229762.1
NM_001242834.1 Intron NP_001229763.1
NM_001242835.1 Intron NP_001229764.1
NM_001242836.1 Intron NP_001229765.1
NM_020465.3 Intron NP_065198.1
NM_022910.3 Intron NP_075061.1

View Full Product Details