Product Details

SNP ID
rs11541056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4672056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGAGCTGGAGGCAGGGTTGGGG[C/G]TGGAGGTGCACCTTCTTCTACCTCC
Phenotype
MIM: 609455
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PELP1 PubMed Links

Gene Details

Gene
PELP1
Gene Name
proline, glutamate and leucine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278241.1 3161 Intron NP_001265170.1
NM_014389.2 3161 Missense Mutation CCC,GCC P1029A NP_055204.3

View Full Product Details