Product Details

SNP ID
rs8111000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53342585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGCCTCATTGTGGAGCCGCTAC[C/T]GTCAGTTCTGTGCCATGGGTCAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF845 PubMed Links

Gene Details

Gene
ZNF845
Gene Name
zinc finger protein 845
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321522.1 Intron NP_001308451.1
NM_001321523.1 Intron NP_001308452.1
NM_001321524.1 Intron NP_001308453.1
NM_138374.2 Intron NP_612383.1

View Full Product Details