Product Details

SNP ID
rs11550994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35869693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGGGTCGGCCCAGGTGGCTGG[A/G]CTATGCGGGCGCCTAACCCTTCACC
Phenotype
MIM: 104775 MIM: 607762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APLP1 PubMed Links

Gene Details

Gene
APLP1
Gene Name
amyloid beta precursor like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024807.2 366 Silent Mutation GGA,GGG G58G NP_001019978.1
NM_005166.4 366 Silent Mutation GGA,GGG G58G NP_005157.1
XM_017026737.1 366 Silent Mutation GGA,GGG G58G XP_016882226.1
XM_017026738.1 366 Silent Mutation GGA,GGG G58G XP_016882227.1
Gene
KIRREL2
Gene Name
kin of IRRE like 2 (Drosophila)
There are no transcripts associated with this gene.

View Full Product Details