Product Details

SNP ID
rs7345556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16273344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATGTTTCGAGAGAGTCAGTCCC[A/C]CTTTGTTGATGTGGAGGGGAGATGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIF16B PubMed Links

Gene Details

Gene
KIF16B
Gene Name
kinesin family member 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199865.1 3894 Missense Mutation GGG,GTG G1237V NP_001186794.1
NM_001199866.1 3894 Intron NP_001186795.1
NM_024704.4 3894 Missense Mutation GGG,GTG G1288V NP_078980.3
XM_005260750.3 3894 Intron XP_005260807.1
XM_005260751.4 3894 Intron XP_005260808.1
XM_005260752.3 3894 Intron XP_005260809.1
XM_005260753.3 3894 Missense Mutation GGG,GTG G1299V XP_005260810.1
XM_005260754.3 3894 Missense Mutation GGG,GTG G1269V XP_005260811.1
XM_005260755.3 3894 Missense Mutation GGG,GTG G1248V XP_005260812.1
XM_006723588.3 3894 Missense Mutation GGG,GTG G1258V XP_006723651.1
XM_017027926.1 3894 Missense Mutation GGG,GTG G1247V XP_016883415.1

View Full Product Details