Product Details

SNP ID
rs6807633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58506153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTAGAATAGTTTTTGGCACAAAG[C/T]ATGTACTTAGTAAATATTTGTTGAA
Phenotype
MIM: 601641
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOX2 PubMed Links

Gene Details

Gene
ACOX2
Gene Name
acyl-CoA oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003500.3 Intron NP_003491.1
XM_005265505.1 Intron XP_005265562.1
XM_006713340.3 Intron XP_006713403.1
XM_017007302.1 Intron XP_016862791.1
Gene
KCTD6
Gene Name
potassium channel tetramerization domain containing 6
There are no transcripts associated with this gene.

View Full Product Details