Product Details
- SNP ID
-
rs9918562
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:57173757 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGAGGCCAGATACCATTCTGAGGC[A/G]CTACAGTAATCACAAACCTACTATC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GUSBP10
PubMed Links
Gene Details
- Gene
- GUSBP10
- Gene Name
- glucuronidase, beta pseudogene 10
There are no transcripts associated with this gene.
View Full Product Details