Product Details

SNP ID
rs28574670
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:2027822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGGAGGAGAAGGGGCCAGAA[A/G]GCCTGGAGCAGTCTGGCTTGTGGGT
Phenotype
MIM: 137163
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRD PubMed Links

Gene Details

Gene
GABRD
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000815.4 Intron NP_000806.2
XM_011541194.2 Intron XP_011539496.1
XM_017000936.1 Intron XP_016856425.1

View Full Product Details