Product Details

SNP ID
rs28373960
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:153545669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACATCTGGATCTCTTTAGCACAT[C/T]TGGCCCCCACCAGCCTCCCCTTCTC
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
There are no transcripts associated with this gene.

Gene
S100A4
Gene Name
S100 calcium binding protein A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002961.2 Intron NP_002952.1
NM_019554.2 Intron NP_062427.1
Gene
S100A5
Gene Name
S100 calcium binding protein A5
There are no transcripts associated with this gene.

Gene
S100A6
Gene Name
S100 calcium binding protein A6
There are no transcripts associated with this gene.

View Full Product Details