Product Details

SNP ID
rs11546377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206160012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAGACCTGAAAAGCCCTGCCGG[C/T]CTCCAGGTGCTCAACGATTACCTGG
Phenotype
MIM: 600655 MIM: 157655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEF1B2 PubMed Links

Gene Details

Gene
EEF1B2
Gene Name
eukaryotic translation elongation factor 1 beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037663.1 419 Intron NP_001032752.1
NM_001959.3 419 Silent Mutation GGC,GGT G11G NP_001950.1
NM_021121.3 419 Intron NP_066944.1
Gene
NDUFS1
Gene Name
NADH:ubiquinone oxidoreductase core subunit S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199981.1 419 Intron NP_001186910.1
NM_001199982.1 419 Intron NP_001186911.1
NM_001199983.1 419 Intron NP_001186912.1
NM_001199984.1 419 Intron NP_001186913.1
NM_005006.6 419 Intron NP_004997.4
XM_017004188.1 419 Intron XP_016859677.1
Gene
SNORA41
Gene Name
small nucleolar RNA, H/ACA box 41
There are no transcripts associated with this gene.

Gene
SNORD51
Gene Name
small nucleolar RNA, C/D box 51
There are no transcripts associated with this gene.

View Full Product Details