Product Details

SNP ID
rs28372691
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218270700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGGTCAGGGCTCTTGGTTCCT[C/G]CCCACACTTTTCTGCAAGATCTTAG
Phenotype
MIM: 603488 MIM: 610147 MIM: 609023 MIM: 610364
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
AAMP PubMed Links

Gene Details

Gene
AAMP
Gene Name
angio associated migratory cell protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001087.4 Intron NP_001078.2
NM_001302545.1 Intron NP_001289474.1
Gene
GPBAR1
Gene Name
G protein-coupled bile acid receptor 1
There are no transcripts associated with this gene.

Gene
MIR6513
Gene Name
microRNA 6513
There are no transcripts associated with this gene.

Gene
PNKD
Gene Name
paroxysmal nonkinesigenic dyskinesia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077399.2 Intron NP_001070867.1
NM_015488.4 Intron NP_056303.3
NM_022572.4 Intron NP_072094.1
XM_017003771.1 Intron XP_016859260.1
XM_017003772.1 Intron XP_016859261.1
Gene
TMBIM1
Gene Name
transmembrane BAX inhibitor motif containing 1
There are no transcripts associated with this gene.

View Full Product Details