Product Details
- SNP ID
-
rs28495769
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:5018641 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCTTCCCCTCTCTGAGTCCCCAG[A/G]TCTGCCTCTGAAGATCGTGCTCACA
- Phenotype
-
MIM: 607930
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYTL1
PubMed Links
Gene Details
- Gene
- CYTL1
- Gene Name
- cytokine like 1
View Full Product Details