Product Details

SNP ID
rs1574473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2852655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATTCAGTGTCTTCTATTTTCT[A/G]TACTAGGTGTTTTCTGTACTGTTTG
Phenotype
MIM: 102680
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADD1 PubMed Links

Gene Details

Gene
ADD1
Gene Name
adducin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001119.4 Intron NP_001110.2
NM_001286645.1 Intron NP_001273574.1
NM_014189.3 Intron NP_054908.2
NM_014190.3 Intron NP_054909.2
NM_176801.2 Intron NP_789771.1
XM_005247933.2 Intron XP_005247990.1
XM_005247934.2 Intron XP_005247991.1
XM_005247935.2 Intron XP_005247992.1
XM_005247936.2 Intron XP_005247993.1
XM_005247937.2 Intron XP_005247994.1
XM_017007703.1 Intron XP_016863192.1
XM_017007704.1 Intron XP_016863193.1
XM_017007705.1 Intron XP_016863194.1
XM_017007706.1 Intron XP_016863195.1
XM_017007707.1 Intron XP_016863196.1
XM_017007708.1 Intron XP_016863197.1
XM_017007709.1 Intron XP_016863198.1
XM_017007710.1 Intron XP_016863199.1

View Full Product Details