Product Details

SNP ID
rs28489605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:26870740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGATCAGTAAGATATTCACCCTA[C/G]CTTGTAGTCTACTAGAAGGCATACC
Phenotype
MIM: 610841
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STIM2 PubMed Links

Gene Details

Gene
STIM2
Gene Name
stromal interaction molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169117.1 Intron NP_001162588.1
NM_001169118.1 Intron NP_001162589.1
NM_020860.3 Intron NP_065911.3

View Full Product Details