Product Details

SNP ID
rs28432037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41834581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGCTTTCTGCTTTCATTTTTCC[A/G]TCCTCATCTTTCTATTACTGGTTGT
Phenotype
MIM: 616786 MIM: 606088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JMJD7 PubMed Links

Gene Details

Gene
JMJD7
Gene Name
jumonji domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114632.1 Intron NP_001108104.1
Gene
JMJD7-PLA2G4B
Gene Name
JMJD7-PLA2G4B readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198588.1 Intron NP_001185517.1
NM_005090.3 Intron NP_005081.1
Gene
MAPKBP1
Gene Name
mitogen-activated protein kinase binding protein 1
There are no transcripts associated with this gene.

Gene
PLA2G4B
Gene Name
phospholipase A2 group IVB
There are no transcripts associated with this gene.

View Full Product Details