Product Details

SNP ID
rs28364552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42412993 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCTAATCCATATTCAATATTAA[A/G]AAAATCAGAAACCAAGGGTGCTGGA
Phenotype
MIM: 114240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN3 PubMed Links

Gene Details

Gene
CAPN3
Gene Name
calpain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000070.2 7882 Intron NP_000061.1
NM_024344.1 7882 Intron NP_077320.1
NM_173087.1 7882 Intron NP_775110.1
NM_173088.1 7882 Intron NP_775111.1
NM_173089.1 7882 Intron NP_775112.1
NM_173090.1 7882 Intron NP_775113.1
Gene
ZNF106
Gene Name
zinc finger protein 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284306.1 7882 UTR 3 NP_001271235.1
NM_001284307.1 7882 UTR 3 NP_001271236.1
NM_022473.2 7882 UTR 3 NP_071918.1
XM_005254591.2 7882 UTR 3 XP_005254648.1
XM_005254592.2 7882 Intron XP_005254649.1
XM_017022483.1 7882 Intron XP_016877972.1
XM_017022484.1 7882 Intron XP_016877973.1
XM_017022485.1 7882 Intron XP_016877974.1

View Full Product Details