Product Details

SNP ID
rs11542097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75019740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATTGTCGGAGTAGTGGTCCTGG[A/G]GGGCAGAATGTGAACAAAGGTACGG
Phenotype
MIM: 603000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL58 PubMed Links

Gene Details

Gene
MRPL58
Gene Name
mitochondrial ribosomal protein L58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303265.1 280 Silent Mutation GGA,GGG G88G NP_001290194.1
NM_001545.2 280 Silent Mutation GGA,GGG G88G NP_001536.1
XM_017024574.1 280 Silent Mutation GGA,GGG G28G XP_016880063.1

View Full Product Details