Product Details
- SNP ID
-
rs28575858
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75463878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTAATGCCACCAAGGATAGAATC[A/C]TACAGCTTGCTATTAAAAACTTCCC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM94
PubMed Links
Gene Details
- Gene
- TMEM94
- Gene Name
- transmembrane protein 94
View Full Product Details