Product Details

SNP ID
rs11537778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4951845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCAGCCATGGCCATGCAGAAA[A/G]TCTTTGCCCGGGAAATCTTGGACTC
Phenotype
MIM: 131370 MIM: 176610 MIM: 610431 MIM: 610056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENO3 PubMed Links

Gene Details

Gene
ENO3
Gene Name
enolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193503.1 540 Intron NP_001180432.1
NM_001976.4 540 Intron NP_001967.3
NM_053013.3 540 Intron NP_443739.3
XM_005256521.2 540 Missense Mutation ATC,GTC I15V XP_005256578.1
XM_011523729.1 540 Missense Mutation ATC,GTC I6V XP_011522031.1
XM_017024346.1 540 Missense Mutation ATC,GTC I6V XP_016879835.1
Gene
PFN1
Gene Name
profilin 1
There are no transcripts associated with this gene.

Gene
RNF167
Gene Name
ring finger protein 167
There are no transcripts associated with this gene.

Gene
SPAG7
Gene Name
sperm associated antigen 7
There are no transcripts associated with this gene.

View Full Product Details