Product Details

SNP ID
hCV60497946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1364967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGAGGCTCTTCTAGCTCCAAT[C/T]ACATTCTTATATGCAACAGATAGGA
Phenotype
MIM: 605066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YWHAE PubMed Links

Gene Details

Gene
YWHAE
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006761.4 Intron NP_006752.1

View Full Product Details