Product Details
- SNP ID
-
rs17883310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:43755022 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACACCGCTCCCTTAAAACCCCAG[A/G]GCGGTGAAAATGCTTCCATTTCTGC
- Phenotype
-
MIM: 605740
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SOST
PubMed Links
Gene Details
- Gene
- SOST
- Gene Name
- sclerostin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025237.2 |
1009 |
UTR 3 |
|
|
NP_079513.1 |
View Full Product Details