Product Details
- SNP ID
-
rs28672974
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:212994867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTCACGAAAATATTTTTCATTAT[A/T]AAAAAAAATTGTTATAAAGTTATTT
- Phenotype
-
MIM: 610471
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANGEL2
PubMed Links
Gene Details
- Gene
- ANGEL2
- Gene Name
- angel homolog 2
- Gene
- VASH2
- Gene Name
- vasohibin 2
There are no transcripts associated with this gene.
View Full Product Details