Product Details
- SNP ID
-
rs3210195
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:84906652 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATTATTTGTATGAATAATGAAA[A/T]TAAAACACACCTCGTGGCATGGCTG
- Phenotype
-
MIM: 188399
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMSB10
PubMed Links
Gene Details
- Gene
- TMSB10
- Gene Name
- thymosin beta 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021103.3 |
|
Intron |
|
|
NP_066926.1 |
View Full Product Details