Product Details

SNP ID
rs1127097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144083804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTCACCGTGCCCTGTGGCTCT[C/G]ACTCTACCAACAGCCAGGCTGTGGG
Phenotype
MIM: 606491 MIM: 603048
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXOSC4 PubMed Links

Gene Details

Gene
EXOSC4
Gene Name
exosome component 4
There are no transcripts associated with this gene.

Gene
GPAA1
Gene Name
glycosylphosphatidylinositol anchor attachment 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003801.3 567 Missense Mutation CAC,GAC H153D NP_003792.1
Gene
MIR6847
Gene Name
microRNA 6847
There are no transcripts associated with this gene.

View Full Product Details