Product Details

SNP ID
rs28548729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143684588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCGCCCCTCCGTCCCTGGGTCGT[A/G]GGCTGTCAAACCTTCTAATTCTGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF707 PubMed Links

Gene Details

Gene
ZNF707
Gene Name
zinc finger protein 707
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100598.1 Intron NP_001094068.1
NM_001100599.1 Intron NP_001094069.1
NM_001288805.1 Intron NP_001275734.1
NM_001288806.1 Intron NP_001275735.1
NM_001288807.1 Intron NP_001275736.1
NM_001288808.1 Intron NP_001275737.1
NM_001288809.1 Intron NP_001275738.1
NM_173831.3 Intron NP_776192.2
XM_005250883.3 Intron XP_005250940.1
XM_006716542.3 Intron XP_006716605.1
XM_006716543.3 Intron XP_006716606.1
XM_011516977.2 Intron XP_011515279.1
XM_011516979.2 Intron XP_011515281.1
XM_017013335.1 Intron XP_016868824.1

View Full Product Details