Product Details

SNP ID
rs28400029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:30877687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTCTGTGTATGGGAACATAGT[A/T]TATGCTTCTGTATCGAGCATATGTG
Phenotype
MIM: 603196
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COCH PubMed Links

Gene Details

Gene
COCH
Gene Name
cochlin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135058.1 651 Silent Mutation GTA,GTT V66V NP_001128530.1
NM_004086.2 651 Silent Mutation GTA,GTT V66V NP_004077.1
XM_017021071.1 651 Silent Mutation GTA,GTT V131V XP_016876560.1
XM_017021072.1 651 Silent Mutation GTA,GTT V131V XP_016876561.1
XM_017021073.1 651 Intron XP_016876562.1
Gene
LOC100506071
Gene Name
uncharacterized LOC100506071
There are no transcripts associated with this gene.

View Full Product Details