Product Details

SNP ID
rs28690138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:46839277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTTAGTATTTTATAAATGCTT[A/T]GTAGTAGGATTGAAACTTTTTCTTA
Phenotype
MIM: 611128
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MDGA2 PubMed Links

Gene Details

Gene
MDGA2
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113498.2 5027 Intron NP_001106970.3
NM_182830.4 5027 Intron NP_878250.2
XM_011536519.2 5027 UTR 3 XP_011534821.1
XM_011536522.2 5027 Intron XP_011534824.1
XM_011536525.2 5027 UTR 3 XP_011534827.1
XM_017021060.1 5027 UTR 3 XP_016876549.1
XM_017021061.1 5027 Intron XP_016876550.1

View Full Product Details