Product Details
- SNP ID
-
rs17845285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:73491672 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGCCCATGCCGCCAGATCACTTT[C/T]ACCGGCGCCTATGGGAGCGCGAGGC
- Phenotype
-
MIM: 614313
MIM: 611919
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACOT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs758109] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACOT1
- Gene Name
- acyl-CoA thioesterase 1
There are no transcripts associated with this gene.
- Gene
- C14orf169
- Gene Name
- chromosome 14 open reading frame 169
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024644.4 |
739 |
Missense Mutation |
CAC,TAC |
H219Y |
NP_078920.2 |
- Gene
- HEATR4
- Gene Name
- HEAT repeat containing 4
View Full Product Details