Product Details

SNP ID
rs29001240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97499380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCCACTGGTGCTGGCGCTTGG[A/G]TTTCCCCGATGGGCTGCTCCGCAGC
Phenotype
MIM: 614777 MIM: 616388
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMS19 PubMed Links

Gene Details

Gene
MMS19
Gene Name
MMS19 homolog, cytosolic iron-sulfur assembly component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289403.1 Intron NP_001276332.1
NM_001289404.1 Intron NP_001276333.1
NM_001289405.1 Intron NP_001276334.1
NM_022362.4 Intron NP_071757.4
XM_005270035.2 Intron XP_005270092.1
XM_005270041.1 Intron XP_005270098.1
XM_006717944.3 Intron XP_006718007.2
XM_006717945.2 Intron XP_006718008.1
XM_011540062.1 Intron XP_011538364.1
XM_011540063.2 Intron XP_011538365.1
XM_017016515.1 Intron XP_016872004.1
XM_017016516.1 Intron XP_016872005.1
XM_017016517.1 Intron XP_016872006.1
XM_017016518.1 Intron XP_016872007.1
XM_017016519.1 Intron XP_016872008.1
XM_017016520.1 Intron XP_016872009.1
XM_017016521.1 Intron XP_016872010.1
XM_017016522.1 Intron XP_016872011.1
XM_017016523.1 Intron XP_016872012.1
XM_017016524.1 Intron XP_016872013.1
XM_017016525.1 Intron XP_016872014.1
XM_017016526.1 Intron XP_016872015.1
XM_017016527.1 Intron XP_016872016.1
XM_017016528.1 Intron XP_016872017.1
XM_017016529.1 Intron XP_016872018.1
XM_017016530.1 Intron XP_016872019.1
XM_017016531.1 Intron XP_016872020.1
XM_017016532.1 Intron XP_016872021.1
XM_017016533.1 Intron XP_016872022.1
XM_017016534.1 Intron XP_016872023.1
XM_017016535.1 Intron XP_016872024.1
Gene
UBTD1
Gene Name
ubiquitin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024954.4 Intron NP_079230.1

View Full Product Details