Product Details

SNP ID
rs28372779
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126269215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGCTCAGAGGGGTTATGATTCG[C/G]AGGGTTCTGCCGCACGGCATGGGCC
Phenotype
MIM: 616587 MIM: 613622 MIM: 182180
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
There are no transcripts associated with this gene.

Gene
FOXRED1
Gene Name
FAD dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017547.3 56 Silent Mutation CGC,CGG R3R NP_060017.1
XM_006718879.3 56 Intron XP_006718942.1
XM_006718881.3 56 UTR 5 XP_006718944.1
XM_011542895.2 56 UTR 5 XP_011541197.1
XM_011542896.2 56 UTR 5 XP_011541198.1
XM_017018000.1 56 Silent Mutation CGC,CGG R3R XP_016873489.1
XM_017018001.1 56 UTR 5 XP_016873490.1
XM_017018002.1 56 UTR 5 XP_016873491.1
XM_017018003.1 56 UTR 5 XP_016873492.1
XM_017018004.1 56 UTR 5 XP_016873493.1
XM_017018005.1 56 Intron XP_016873494.1
XM_017018006.1 56 UTR 5 XP_016873495.1
Gene
SRPRA
Gene Name
SRP receptor alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177842.1 56 Intron NP_001171313.1
NM_003139.3 56 Intron NP_003130.2
XM_017018179.1 56 Intron XP_016873668.1

View Full Product Details