Product Details

SNP ID
rs17878948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102320480 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCAAGTCACCCAGTTTGAGGCTG[G/T]TCTCAAACACCTGGGCTCAAGCAAT
Phenotype
MIM: 601721
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BIRC3 PubMed Links

Gene Details

Gene
BIRC3
Gene Name
baculoviral IAP repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165.4 Intron NP_001156.1
NM_182962.2 Intron NP_892007.1
XM_017017643.1 Intron XP_016873132.1

View Full Product Details