Product Details

SNP ID
rs74050546
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:3490501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTCCCCTCTGGCTGGGACTGGA[G/T]AGGCGGGCTCCACGGGACTGCCTCT
Phenotype
MIM: 604266
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF16 PubMed Links

Gene Details

Gene
ARHGEF16
Gene Name
Rho guanine nucleotide exchange factor 16
There are no transcripts associated with this gene.

Gene
MEGF6
Gene Name
multiple EGF like domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001409.3 4515 UTR 3 NP_001400.3
XM_006710406.3 4515 UTR 3 XP_006710469.1
XM_011540885.1 4515 UTR 3 XP_011539187.1
XM_011540886.1 4515 UTR 3 XP_011539188.1
XM_011540887.2 4515 UTR 3 XP_011539189.1
XM_011540888.2 4515 Intron XP_011539190.1
XM_017000533.1 4515 UTR 3 XP_016856022.1

View Full Product Details