Product Details

SNP ID
rs61745042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31792130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTTTGCGGGGGGCTTTCCCAC[C/T]GGTGGATTTACGAGCAGTCTGCTTG
Phenotype
MIM: 616134
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H3F3C PubMed Links

Gene Details

Gene
H3F3C
Gene Name
H3 histone, family 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013699.2 Intron NP_001013721.2

View Full Product Details