Product Details

SNP ID
rs117499206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40740983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAGGCTGGTTCACTCACTTGG[C/G]AGTCACTGGGCGGCTATTCTTGTGC
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 2166 UTR 3 NP_940878.3
XM_005258776.4 2166 UTR 3 XP_005258833.1
XM_005258778.4 2166 UTR 3 XP_005258835.1
XM_006723152.2 2166 Intron XP_006723215.1
XM_006723153.2 2166 Intron XP_006723216.1
XM_011526774.2 2166 UTR 3 XP_011525076.1
XM_011526775.1 2166 Intron XP_011525077.1
XM_011526776.2 2166 UTR 3 XP_011525078.1
XM_011526777.2 2166 UTR 3 XP_011525079.1
XM_011526778.2 2166 UTR 3 XP_011525080.1
XM_011526779.2 2166 UTR 3 XP_011525081.1
XM_011526782.2 2166 Intron XP_011525084.1
XM_011526783.2 2166 UTR 3 XP_011525085.1
XM_011526784.2 2166 UTR 3 XP_011525086.1
XM_011526785.2 2166 UTR 3 XP_011525087.1
XM_011526786.2 2166 Intron XP_011525088.1
XM_017026620.1 2166 UTR 3 XP_016882109.1
XM_017026621.1 2166 UTR 3 XP_016882110.1
XM_017026622.1 2166 UTR 3 XP_016882111.1
XM_017026623.1 2166 UTR 3 XP_016882112.1
XM_017026624.1 2166 UTR 3 XP_016882113.1
XM_017026625.1 2166 Intron XP_016882114.1
XM_017026626.1 2166 UTR 3 XP_016882115.1
XM_017026627.1 2166 UTR 3 XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025194.2 2166 Intron NP_079470.1
XM_017027324.1 2166 Intron XP_016882813.1
Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details