Product Details

SNP ID
rs61734464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27494021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGGACTCTCTCCCCGAAGGCC[G/T]ATGCTCCTGACCTGCACTGTGTAGT
Phenotype
MIM: 611905 MIM: 600842 MIM: 607386
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FNDC4 PubMed Links

Gene Details

Gene
FNDC4
Gene Name
fibronectin type III domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022823.2 769 Silent Mutation NP_073734.1
XM_005264499.3 769 Silent Mutation XP_005264556.1
Gene
GCKR
Gene Name
glucokinase (hexokinase 4) regulator
There are no transcripts associated with this gene.

Gene
IFT172
Gene Name
intraflagellar transport 172
There are no transcripts associated with this gene.

View Full Product Details