Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_138694.3 | 12247 | UTR 3 | NP_619639.3 | ||
NM_170724.2 | 12247 | Intron | NP_733842.2 | ||
XM_011514680.2 | 12247 | UTR 3 | XP_011512982.1 | ||
XM_011514682.2 | 12247 | UTR 3 | XP_011512984.1 | ||
XM_011514683.2 | 12247 | UTR 3 | XP_011512985.1 | ||
XM_011514684.2 | 12247 | UTR 3 | XP_011512986.1 | ||
XM_011514685.1 | 12247 | Intron | XP_011512987.1 | ||
XM_011514686.2 | 12247 | Intron | XP_011512988.1 | ||
XM_011514687.1 | 12247 | Intron | XP_011512989.1 | ||
XM_011514688.2 | 12247 | Intron | XP_011512990.1 | ||
XM_011514690.2 | 12247 | UTR 3 | XP_011512992.1 | ||
XM_011514691.2 | 12247 | UTR 3 | XP_011512993.1 | ||
XM_017010944.1 | 12247 | UTR 3 | XP_016866433.1 | ||
XM_017010945.1 | 12247 | UTR 3 | XP_016866434.1 | ||
XM_017010946.1 | 12247 | UTR 3 | XP_016866435.1 | ||
XM_017010947.1 | 12247 | UTR 3 | XP_016866436.1 | ||
XM_017010948.1 | 12247 | UTR 3 | XP_016866437.1 | ||
XM_017010949.1 | 12247 | UTR 3 | XP_016866438.1 | ||
XM_017010950.1 | 12247 | Intron | XP_016866439.1 | ||
XM_017010951.1 | 12247 | Intron | XP_016866440.1 | ||
XM_017010952.1 | 12247 | Intron | XP_016866441.1 |