Product Details

SNP ID
rs41307375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:66022315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAACTCATAATCCAGAGGAACT[A/G]TGTCCAGCAGGCGGGCGTAGTTGCC
Phenotype
MIM: 300694 MIM: 300353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR223 PubMed Links

Gene Details

Gene
MIR223
Gene Name
microRNA 223
There are no transcripts associated with this gene.

Gene
VSIG4
Gene Name
V-set and immunoglobulin domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100431.1 1275 Missense Mutation ACA,ATA T289I NP_001093901.1
NM_001184830.1 1275 UTR 3 NP_001171759.1
NM_001184831.1 1275 UTR 3 NP_001171760.1
NM_001257403.1 1275 Intron NP_001244332.1
NM_007268.2 1275 Missense Mutation ACA,ATA T383I NP_009199.1
XM_017029251.1 1275 Intron XP_016884740.1

View Full Product Details