Product Details

SNP ID
rs17421067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183510790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGTCTTATGGGGAGATGATAGG[A/C]TGTGACAATGAACAGTGTCCAATTG
Phenotype
MIM: 604215
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ING2 PubMed Links

Gene Details

Gene
ING2
Gene Name
inhibitor of growth family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291959.1 672 Silent Mutation GGA,GGC G187G NP_001278888.1
NM_001564.3 672 Silent Mutation GGA,GGC G227G NP_001555.1
XM_011531927.2 672 Silent Mutation GGA,GGC G172G XP_011530229.1
Gene
LOC389247
Gene Name
uncharacterized LOC389247
There are no transcripts associated with this gene.

View Full Product Details