Product Details

SNP ID
rs61828274
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:230984459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAAAGGAAGTTGCAAGCAGCAC[G/T]TAAGGGAAAAGCAAATAATAGAGGG
Phenotype
MIM: 611647
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARV1 PubMed Links

Gene Details

Gene
ARV1
Gene Name
ARV1 homolog, fatty acid homeostasis modulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022786.1 Intron NP_073623.1
XM_017002106.1 Intron XP_016857595.1
Gene
TTC13
Gene Name
tetratricopeptide repeat domain 13
There are no transcripts associated with this gene.

View Full Product Details