Product Details

SNP ID
rs61755260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:149575763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTACTTCTTCACTCCAAACAGTC[A/G]TATCATTCTTAGTTTTTTGTGTTAC
Phenotype
MIM: 611935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMADHC PubMed Links

Gene Details

Gene
MMADHC
Gene Name
methylmalonic aciduria and homocystinuria, cblD type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015702.2 Intron NP_056517.1

View Full Product Details