Product Details

SNP ID
rs56264174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5016917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTACCTGGGCCACTTTCCAACA[C/T]GGGGGCGAGGCCACAAAGTCCCGCA
Phenotype
MIM: 607930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYTL1 PubMed Links

Gene Details

Gene
CYTL1
Gene Name
cytokine like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018659.2 271 Silent Mutation CCA,CCG P82P NP_061129.1
XM_017008299.1 271 Missense Mutation ATG,GTG M121V XP_016863788.1

View Full Product Details