Product Details

SNP ID
rs59037741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17843847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACACATTTAACAAGTAACATTT[C/T]TAGGGAAAATGAAGGAAGTACCACA
Phenotype
MIM: 611799 MIM: 606280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCORL PubMed Links

Gene Details

Gene
LCORL
Gene Name
ligand dependent nuclear receptor corepressor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166139.2 7686 Intron NP_001159611.1
NM_153686.8 7686 UTR 3 NP_710153.2
XM_011513821.2 7686 UTR 3 XP_011512123.1
XM_011513822.2 7686 UTR 3 XP_011512124.1
XM_011513823.2 7686 UTR 3 XP_011512125.1
XM_011513825.2 7686 Intron XP_011512127.1
XM_011513826.2 7686 UTR 3 XP_011512128.1
XM_017007961.1 7686 UTR 3 XP_016863450.1
XM_017007962.1 7686 UTR 3 XP_016863451.1
XM_017007963.1 7686 UTR 3 XP_016863452.1
XM_017007964.1 7686 Intron XP_016863453.1
XM_017007965.1 7686 Intron XP_016863454.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022346.4 7686 UTR 3 NP_071741.2
XM_011513876.2 7686 UTR 3 XP_011512178.1
XM_017008543.1 7686 UTR 3 XP_016864032.1
XM_017008544.1 7686 UTR 3 XP_016864033.1

View Full Product Details