Product Details
- SNP ID
-
rs56919691
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:166306678 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGTTTCAGGGATGTCTAATACC[C/T]TCAACATTTTTCTGACTTGGGAGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRR18
PubMed Links
Gene Details
- Gene
- PRR18
- Gene Name
- proline rich 18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175922.3 |
1706 |
UTR 3 |
|
|
NP_787118.2 |
View Full Product Details