Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001122847.2 | 3444 | UTR 3 | NP_001116319.1 | ||
NM_001122848.2 | 3444 | UTR 3 | NP_001116320.1 | ||
NM_001206931.1 | 3444 | UTR 3 | NP_001193860.1 | ||
NM_003044.4 | 3444 | UTR 3 | NP_003035.3 | ||
XM_005253747.4 | 3444 | Intron | XP_005253804.1 | ||
XM_005253748.4 | 3444 | UTR 3 | XP_005253805.1 | ||
XM_006719005.3 | 3444 | Intron | XP_006719068.1 | ||
XM_011521010.1 | 3444 | UTR 3 | XP_011519312.1 | ||
XM_017019841.1 | 3444 | UTR 3 | XP_016875330.1 |