Product Details

SNP ID
rs62058606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30921078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGCGGCAAAGAAGCCTCCTA[C/T]CCAGCCTTGGGCAAGTCTCTGCCAA
Phenotype
MIM: 609085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL19 PubMed Links

Gene Details

Gene
FBXL19
Gene Name
F-box and leucine rich repeat protein 19
There are no transcripts associated with this gene.

Gene
FBXL19-AS1
Gene Name
FBXL19 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details